Would you like to log in again?
Conical Centrifuge Tube
Supplier: EISCO SCIENTIFIC LLC
CH0268CPK12
470345-072EA 24.89 USD InStock
470345-072
Conical Centrifuge Tube
Tubes Centrifuge Tubes
Pack of 12 conical centrifuge tubes made of borosilicate glass with a 15 ml capacity, measuring 120×17 mm (OD), providing durability and accurate sample handling.
  • Twelve glass conical centrifuge tubes with 17 mm diameter and 15 ml capacity
  • Tubes are ideal for centrifugation as well as sample storage and culture growth
  • Conical shape encourages material mixing within tube
  • Economic package size can fill classroom needs effectively
  • Appropriate in professional and home lab settings
A pack of 12 centrifuge tubes with 15 ml capacity. Ideal for use in a centrifuge, these tubes may also be used for sample handling and storage, as well as culturing. The versatility in their function makes them perfect for classroom lab settings, and the lab-quality borosilicate glass makes them a perfect supply for professional labs as well. Autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures and thermal shocks. Eisco Borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty