Would you like to log in again?
Gooch Crucibles
Supplier: EISCO SCIENTIFIC LLC
CH0509EPK5
470345-938EA 19.15 USD InStock
470345-938
Gooch Crucibles
Crucibles
Set of 5 glazed porcelain Gooch crucibles with a 30 ml capacity, featuring an unglazed rim and bottom surface, ensuring accurate filtration and sample preparation for various laboratory applications.
  • Crucibles are made of porcelain and have a perforated bottom
  • Each crucible is glazed inside and outside with the exception of outside bottom surface and rim
  • Top outside diameter 42 mm, overall height 40 mm, 2 mm thickness
  • 30 ml capacity
Pack of 5, 30 ml capacity porcelain filtering crucibles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty