Would you like to log in again?
Wooden Pipette Rack
Supplier: EISCO SCIENTIFIC LLC
CH0590A
470343-702EA 50.7 USD InStock
470343-702
Wooden Pipette Rack
Pipette Stands
Polished wood construction pipette rack designed to hold 24 pipettes vertically, ensuring easy access and efficient organization of pipettes.
  • Premium quality pipette stand featuring a sturdy wood construction that will stand up to the wear and tear of the lab
  • Rack holds pipettes vertically, allowing liquids to drain. Measures 15.5" wide, 3.25" long, and 16.25" tall, allowing it to fit in most lab spaces
  • Supports up to 24 pipettes - rack features (8) 20 mm holes, (8) 12 mm holes and (8) 9 mm holes. Can hold pipettes with diameters from 0.9 to 2 cm
  • Designed for use in professional laboratories, but is an ideal choice for science classrooms
Premium quality wooden rack designed to hold up to 24 serological pipettes. Polished. Rack features holes measuring 9, 12 and 20 mm in diameter to accommodate an assortment of pipettes. Rack holds pipettes straight and vertical, allowing liquids to drain and labware to dry. Measures 15.5" wide, 3.25" long, and 16.25" tall. Designed for use in professional laboratories, but is an ideal choice for science classrooms. The design of this rack allows it to be repurposed in a variety of ways - can be used in arts and crafts to store and dry paint brushes or tools.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty