Would you like to log in again?
Rotary Pipette Stand
Supplier: EISCO SCIENTIFIC LLC
CH0595
470345-190EA 24.9 USD InStock
470345-190
Rotary Pipette Stand
Pipette Stands
Rotary pipette stand capable of holding 94 pipettes vertically, featuring an open bottom for easy draining, allowing for convenient and organized storage of pipettes.
  • Pipette stand featuring a sturdy polypropylene construction that will stand up to the wear and tear of the lab
  • Stand can hold maximum of 94 pipettes and rotate on a vertical axis
  • The lower rotating disc has four concentric troughs with tapered sides and open bottoms which create the pipettes tape protectively and permit water drops to run off
  • Designed for use in professional laboratories, but is an ideal choice for science classroom
Polypropylene rotary pipette stand, Base is 9" (228 mm) Diameter and measures around 19" (482 mm) tall. Holds 94 pipettes. This will hold a number of variations of sizes of pipettes. Max size diameter pipette to be stored would 0.50" (12.7 mm) or less. Autoclavable and chemical resistant. Glass Pipettes are not include with stand.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty