Would you like to log in again?
Wooden Pipette Rack
Supplier: EISCO SCIENTIFIC LLC
CH0590B
470344-850EA 38.05 USD InStock
470344-850
Wooden Pipette Rack
Pipette Stands
Unpolished wood construction pipette rack with a capacity to hold 12 pipettes vertically, providing a cost-effective and functional storage option for pipettes.
  • Premium quality pipette stand featuring a sturdy wood construction that will stand up to the wear and tear of the lab
  • Rack holds pipettes vertically, allowing liquids to drain
  • Two legs extend backward to angle the rack slightly
  • Measures 12" wide and 14.75" tall, allowing it to fit in most lab spaces
  • Supports up to 12 pipettes
  • Designed for use in professional laboratories, but is an ideal choice for science classrooms
Rack features a variety of slots to accommodate an assortment of pipette sizes - (4) 20 mm, (2) 14 mm, (2) 12.5 mm and (4) 10 mm slots. Fits pipettes with diameters ranging from 0.9 to 2 cm. Rack holds pipettes vertical, at a slight angle, allowing liquids to drain and pipettes to dry. Measures 12" wide, 5.75" long (when legs are extended) and 14.75" tall. The design of this rack allows it to be repurposed in a variety of ways - can be used in arts and crafts to store and dry paint brushes or tools.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty