Would you like to log in again?
Clamp Retort
Supplier: EISCO SCIENTIFIC LLC
CH0687
470344-136EA 17.39 USD InStock
470344-136
Clamp Retort
Clamps General Purpose Clamps
Retort clamp with 4 prongs, cork-lined jaws, and a boss head for attaching to rods up to 15 mm, offering secure and adjustable holding for objects up to 90 mm in laboratory setups and experiments.
  • Cork lined jaws
  • Perfect for test tubes, burettes, or flasks
Four prong, die casted alloy clamp retort with cork lined jaws with boss head.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty