Would you like to log in again?
Plated Spring Steel Wire Test Tube Clamps with Finger Grips
Supplier: EISCO SCIENTIFIC LLC
CH073010
470343-582EA 20.8 USD InStock
470343-582
Plated Spring Steel Wire Test Tube Clamps with Finger Grips
Clamps General Purpose Clamps
With finger grips, this pack of 10 clamps ensures a secure hold on test tubes, offering convenience and reliability in laboratory experiments.
  • Pack of 10 quality plated steel wire test tube holders
  • Finger grips make use comfortable and secure
  • Self closing clamp for ease of use
  • Lightweight, Space efficient
  • Durable enough for student use in the classroom or lab
Pack of ten wire test tube clamps. Economically priced, maximally functional plated spring steel wire holders with finger grips. The jaws will accept tubes up to 40 mm diameter and their alignment is maintained by a wire collar. Overall length 6.5".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty