Would you like to log in again?
Thermo Scientific™ Piercing Plate for Matrix™ 2D Barcoded Storage Tubes, 384
Supplier: Thermo Scientific
Matrix®
4960
76642-936CS 154.69 USD InStock
76642-936
Thermo Scientific™ Piercing Plate for Matrix™ 2D Barcoded Storage Tubes, 384
Microplates Deep Well Microplates
Certifications: Products are manufactured in ISO 7 class clean room environment according to ISO 14644-1:2015.
Sterile products are sterilized by gamma irradiation according to ISO 11137; 10⁻³ Sterility Assurance Level (SAL)
The vials and closures were tested and met the requirements of USP <88> Class VI plastics.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty