Would you like to log in again?
Flexible Mirrors
Supplier: United Scientific Supplies
MSAST6
470339-376EA 37.4 USD InStock
470339-376
Flexible Mirrors
Mirrors
Flexible mirrors for use in optical experiments.
  • Acts as a concave or convex mirror
  • Can be cut with scissors
These acrylic double-sided mirrors are flexible and can be used for demonstrating concave and convex properties. Material can be cut easily with scissors. This set contains four 7.5×6" mirrors, four 4×3" mirrors and four 3×2" mirrors. Mirror supports not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty