Would you like to log in again?
Vernier Friction Pad DTS
Supplier: VERNIER SOFTWARE & TECH SE
Vernier
DTS-PAD
470319-984EA 31.99 USD InStock
470319-984
Vernier Friction Pad DTS
Educational Materials Physics Educational Materials Mechanics Learning Activities
Add a Friction Pad DTS to your Vernier Dynamics Cart to produce a very consistent friction force.
  • Compatible with Go Direct® Sensor Cart, Standard Cart, Plunger Cart, or Motion Encoder Cart
  • Study friction in a controlled and adjustable way
After working hard to make our dynamics carts very low friction, we’ve added some friction back. Now you and your students can study friction in a controlled way.
Add a Friction Pad DTS to your Vernier Go Direct Sensor Cart, Standard Cart, Plunger Cart, or Motion Encoder Cart, and watch the carts move under very consistent friction force. Setting the friction amount is as simple as turning a knob.
Note: This Friction Pad is not compatible with our older metal dynamics carts that were part of our Vernier Dynamics Systems that were sold from 2005 to 2012.
Ordering information: Vernier dynamics cart required but not included.
Delivery information: Includes 1 friction pad.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty