Would you like to log in again?
Vernier Go Direct® Fluorescence/UV-VIS Spectrophotometer
Supplier: VERNIER SOFTWARE & TECH SE
Vernier
GDX-SPEC-FUV
470338-802EA 2998.99 USD InStock
470338-802
Vernier Go Direct® Fluorescence/UV-VIS Spectrophotometer
Spectroscopic Instruments Fluorescence Spectrophotometers
Measure fluorescence and absorbance spectra of myriad samples.
  • Compatible with LabQuest® 2 and 3, and computers running the Spectral Analysis® app
  • Optical resolution 3.0 nm
Set your students up for success with the Go Direct® Fluorescence/UV-VIS Spectrophotometer. Geared toward upper-level chemistry, this innovative device combines the functionality of a fluorometer with the power of a UV/VIS spectrophotometer. The Go Direct Fluorescence/UV-VIS Spectrophotometer allows students to conduct fluorescence data collection back-to-back with absorbance data collection in a single instrument.
This supports quantum yield studies, quenching investigations, kinetics, Beer’s law, and simple fluorescence detection with samples such as quinine sulfate, DAPI, GFP, and tryptophan. Excitation LEDs included are 375 nm, 450 nm, and 525 nm—other wavelengths can be purchased separately. Inquire about custom wavelengths. To conduct emission spectra experiments, simply attach the Vernier Spectrophotometer Optical Fiber.
Additionally, the Go Direct® Fluorescence/UV-VIS Spectrophotometer connects directly to a tablet, computer, or LabQuest®, offering students the flexibility to work with many technologies used in chemistry labs. Data can be collected and analyzed using our LabQuest® 2, LabQuest® 3, Logger Pro® software, or free Vernier Spectral Analysis® software, which allows users to spend less time learning different interfaces and more time in the laboratory.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty