Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Vernier Go Direct® Emissions Spectrometer
Supplier: VERNIER SOFTWARE & TECH SE
Vernier
GDX-SPEC-EM 
470338-796EA 949.99 USD InStock
470338-796
Vernier Go Direct® Emissions Spectrometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Measure and examine emissions spectra of light bulbs, spectrum tubes, or the sun.
  • Compatible with LabQuest® 2 and 3, and computers running the Spectral Analysis® app
Elevate emissions analysis with the Go Direct® emissions spectrometer. This device connects via Bluetooth® wireless technology or USB to a tablet, computer, or LabQuest®. With the Go Direct® emissions spectrometer, students can measure spectra of light-emitting samples in the range of 350 to 950 nm. The sensitivity of the linear CCD allows this spectrometer to be used with light bulbs, spectrum tubes, or the sun.
Display and analyze spectral data on LabQuest® 2, LabQuest® 3, Logger Pro®, or Vernier Spectral Analysis®. For analyzing the emissions of a spectrum tube, an optical fiber is recommended. For measuring spectra of large sources (such as the sun) or comparing types of light bulbs, the optical fiber is not necessary.
Ordering information: Vernier emissions fiber cable is recommended (sold separately).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty