Would you like to log in again?
EnGen® Spy Cas9 HF1
Supplier: New England Biolabs
M0667T M0667M
76717-054EA 724 USD InStock
76717-054 76717-036
EnGen® Spy Cas9 HF1
Nucleic Acid Reagents Cloning Kits and Ligation
EnGen® Spy Cas9 HF1 is a high-fidelity, quadruple substitution (N497A/R661A/Q695A/Q926A) variant of EnGen Spy Cas9 NLS from streptococcus pyogenes with reduced non-specific DNA cleavage.
  • High-fidelity variant of Spy Cas9 nuclease reduces off-target cleavage
  • Ideal for direct introduction of Cas9/sgRNA complexes
  • Compatible with EnGen sgRNA synthesis Kit, S. pyogenes and the EnGen mutation detection kit
Spy Cas9 is an RNA-guided endonuclease that catalyzes site-specific cleavage of double stranded DNA. The single guide RNA (sgRNA) targets Cas9 to the region immediately upstream of a 5'-NGG-3' protospacer adjacent motif (PAM) producing a double stranded break 3 bases upstream of the PAM.
EnGen Spy Cas9 HF1 contains Simian virus 40 (SV40) T antigen nuclear localization sequence (NLS) on the N- and C-termini of the protein.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty