Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver(I) hexafluorophosphate 98%

Supplier: Ambeed
Danger
Synonyms: Silver hexafluorophosphate

A129287-1G A129287-25G A129287-5G
76718-892EA 44.29 USD InStock
76718-892 76718-894 76718-896
Silver(I) hexafluorophosphate 98%
Silver (I) hexafluorophosphate
Caution: for Research Use Only


Formula: AgF₆P
MW: 252.83 g/mol
MDL Number: MFCD00003415
CAS Number: 26042-63-7
UN: 3260
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty