Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propyl benzoate 99%

Supplier: Ambeed
Warning
Synonyms: Benzoic acid propyl ester

A131383-10G A131383-100G A131383-25G
76718-970EA 9.27 USD InStock
76718-970 76718-972 76718-968
Propyl benzoate 99%
Propyl benzoate
Caution: for Research Use Only


Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00009370
CAS Number: 2315-68-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty