Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pentadecanoic acid 98%

Supplier: Ambeed
Warning
Synonyms: Pentadecylic acidn-Pentadecanoic acid

A135858-5G A135858-100G
76719-176EA 17.51 USD InStock
76719-176 76719-174
Pentadecanoic acid 98%
Pentadecanoic acid
Caution: for Research Use Only


Formula: C₁₅H₃₀O₂
MW: 242.4 g/mol
MDL Number: MFCD00002745
CAS Number: 1002-84-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty