Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ɑ-Asarone 95%

Supplier: Ambeed
Warning
Synonyms: trans-1,2,4-Trimethoxy-5-(1-propenyl)benzene(E)-1,2,4-Trimethoxy-5-(prop-1-en-1-yl)benzene

A154546-1G A154546-1MG
76719-880EA 13.39 USD InStock
76719-880 76719-882
ɑ-Asarone 95%
alpha-Asarone
Caution: for Research Use Only


Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
MDL Number: MFCD00064457
CAS Number: 2883-98-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty