Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxamic acid 95%

Supplier: Ambeed
Danger
Synonyms: 2-Amino-2-oxoacetic acid

A178440-1G A178440-25G A178440-5G
76720-788EA 23.69 USD InStock
76720-788 76720-786 76720-784
Oxamic acid 95%
Oxamic acid
Caution: for Research Use Only


Formula: C₂H₃NO₃
MW: 89.05 g/mol
MDL Number: MFCD00008006
CAS Number: 471-47-6
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty