Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

p-Toluamide 97%

Supplier: Ambeed
Warning
Synonyms: 4-Methylbenzamide

A193666-5G A193666-100G A193666-10G A193666-25G
76721-192EA 57.68 USD InStock
76721-192 76721-194 76721-196 76721-198
p-Toluamide 97%
p-Toluamide
Caution: for Research Use Only


Formula: C₈H₉NO
MW: 135.17 g/mol
MDL Number: MFCD00008002
CAS Number: 619-55-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty