Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitrosobenzene 98%

Supplier: Ambeed
Danger

A195460-5G A195460-1G
76721-270EA 6.18 USD InStock
76721-270 76721-272
Nitrosobenzene 98%
Nitrosobenzene
Caution: for Research Use Only


Formula: C₆H₅NO
MW: 107.11 g/mol
MDL Number: MFCD00002059
CAS Number: 586-96-9
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty