Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Diethylacetamide 98%

Supplier: Ambeed
Warning
Synonyms: N-Acetyldiethylamine

A290507-10G A290507-25G A290507-5G A290507-100G A290507-500G
76723-500EA 5.15 USD InStock
76723-500 76723-492 76723-494 76723-496 76723-498
N,N-Diethylacetamide 98%
N,N-Diethylacetamide
Caution: for Research Use Only


Formula: C₆H₁₃NO
MW: 115.18 g/mol
MDL Number: MFCD00009047
CAS Number: 685-91-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty