Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nabumetone 98%

Supplier: Ambeed
Warning
Synonyms: 4-(6-Methoxynaphthalen-2-yl)butan-2-one

A307752-1MG A307752-10MG A307752-50MG
76723-774EA 5.15 USD InStock
76723-774 76723-776 76723-772
Nabumetone 98%
Nabumetone
Caution: for Research Use Only


Formula: C₁₅H₁₆O₂
MW: 228.29 g/mol
MDL Number: MFCD00079518
CAS Number: 42924-53-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty