Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Phenoxynicotinic acid 98%

Supplier: Ambeed
Warning

A331090-100G A331090-25G A331090-5G A331090-1G
76724-174EA 32.96 USD InStock
76724-174 76724-172 76724-170 76724-168
2-Phenoxynicotinic acid 98%
2-Phenoxynicotinic acid
Caution: for Research Use Only


Formula: C₁₂H₉NO₃
MDL Number: MFCD00014629
CAS Number: 35620-71-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty