Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phthalic acid 98%

Supplier: Ambeed
Warning
Synonyms: Pathalic acid

A352709-500G A352709-25G A352709-100G
76724-486EA 17.51 USD InStock
76724-486 76724-484 76724-482
Phthalic acid 98%
Phthalic acid
Caution: for Research Use Only


Formula: C₈H₆O₄
MW: 166.13 g/mol
MDL Number: MFCD00002467
CAS Number: 88-99-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty