Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium carbonate 99%

Supplier: Ambeed
Warning
Synonyms: Carbonic acid potassium saltCarbonic acid, dipotassium salt

A442495-1000G A442495-100G A442495-500G
76725-482EA 11.33 USD InStock
76725-482 76725-484 76725-486
Potassium carbonate 99%
Potassium carbonate
Caution: for Research Use Only


Formula: K₂CO₃
MW: 138.21 g/mol
MDL Number: MFCD00011382
CAS Number: 584-08-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty