Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perfluorotoluene 98%

Supplier: Ambeed
Danger
Synonyms: Octafluorotoluene1,2,3,4,5-Pentafluoro-6-(trifluoromethyl)benzeneα,α,α,2,3,4,5,6-OctafluorotoluenePentafluoro(trifluoromethyl)benzene

A464630-5G A464630-25G
76725-726EA 16.48 USD InStock
76725-726 77551-412
Perfluorotoluene 98%
Perfluorotoluene
Caution: for Research Use Only


Formula: C₇F₈
MW: 236.06 g/mol
MDL Number: MFCD00000375
CAS Number: 434-64-0
UN: 1992
ADR: 3,6.1,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty