Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biphenyl diphenyl ether eutectic 26.5%diphenyl+73.5%diphenyl oxide

Supplier: Ambeed
Danger
Synonyms: Dowtherm® ADiphyl

A493059-25G
76725-976EA 6.18 USD InStock
76725-976
Biphenyl diphenyl ether eutectic 26.5%diphenyl+73.5%diphenyl oxide
Biphenyl diphenyl ether eutectic
Caution: for Research Use Only


Formula: C₂₄H₂₀O
MW: 324.423 g/mol
MDL Number: MFCD00148859
CAS Number: 8004-13-5
UN: 3082
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty