Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyltrimethoxysilane 98%

Supplier: Ambeed
Danger
Synonyms: Trimethoxy(methyl)silane

A506558-25G
76726-080EA 6.18 USD InStock
76726-080
Methyltrimethoxysilane 98%
Methyltrimethoxysilane
Caution: for Research Use Only


Formula: C₄H₁₂O₃Si
MDL Number: MFCD00008342
CAS Number: 1185-55-3
UN: 1993
ADR: 3,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty