Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-5-amino-2-furoate 98%

Supplier: Ambeed
Warning
Synonyms: Methyl 5-aminofuran-2-carboxylate

A522818-1G A522818-250MG A522818-100MG
76726-218EA 29.87 USD InStock
76726-218 76726-216 76726-214
Methyl-5-amino-2-furoate 98%
Methyl 5-amino-2-furoate
Caution: for Research Use Only


Formula: C₆H₇NO₃
MW: 141.13 g/mol
MDL Number: MFCD00051661
CAS Number: 22600-30-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty