Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Dodecylmethylamine 95%

Supplier: Ambeed
Danger
Synonyms: N-methyldodecylamine

A546067-5G A546067-1G A546067-250MG
76726-428EA 181.28 USD InStock
76726-428 76726-426 76726-424
N-Dodecylmethylamine 95%
N-Dodecylmethylamine
Caution: for Research Use Only


Formula: CH₃(CH₂)₁₁NHCH₃
MW: 199.38 g/mol
MDL Number: MFCD00026574
CAS Number: 7311-30-0
UN: 2735
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty