Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Piperidine-1-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A637744-5G A637744-1G
76727-208EA 16.48 USD InStock
76727-208 76727-210
Piperidine-1-sulfonyl chloride 95%
Piperidine-1-sulfonyl chloride
Caution: for Research Use Only


Formula: C₅H₁₀ClNO₂S
MW: 183.65 g/mol
MDL Number: MFCD03250329
CAS Number: 35856-62-3
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty