Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylacetamide 98%

Supplier: Ambeed
Danger
Synonyms: Acetic acid dimethylamideDimethyl Acetamide

A705242-25G A705242-500G A705242-10G
76727-724EA 6.18 USD InStock
76727-724 76727-726 76727-722
N,N-Dimethylacetamide 98%
N,N-Dimethylacetamide
Caution: for Research Use Only


Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
MDL Number: MFCD00008686
CAS Number: 127-19-5
UN: 3082
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty