Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Octylthiophene 97%

Supplier: Ambeed
Warning
Synonyms: 2-n-Octylthiophene

A765994-1G
76728-226EA 15.45 USD InStock
76728-226
2-Octylthiophene 97%
2-Octylthiophene
Caution: for Research Use Only


Formula: C₁₂H₂₀S
MW: 196.36 g/mol
MDL Number: MFCD00041015
CAS Number: 880-36-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty