Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-DL-leucine 98%

Supplier: Ambeed
Warning
Synonyms: 2-Acetamido-4-methylpentanoic acidAc-DL-Leu-OH

A772477-100G A772477-25G
76728-274EA 7.21 USD InStock
76728-274 76728-272
N-Acetyl-DL-leucine 98%
N-Acetyl-DL-leucine
Caution: for Research Use Only


Formula: C₈H₁₅NO₃
MW: 173.2 g/mol
MDL Number: MFCD00026498
CAS Number: 99-15-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty