Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitrilotriacetic acid trisodium salt monohydrate 98%

Supplier: Ambeed
Danger
Synonyms: Nitrilotriacetic acid, trisodium salt, monohydrate

A820906-25G
76728-694EA 5.15 USD InStock
76728-694
Nitrilotriacetic acid trisodium salt monohydrate 98%
Nitrilotriacetic acid trisodium salt monohydrate
Caution: for Research Use Only


Formula: C₆H₆NNa₃O₆·1H₂O
MW: 275.1 g/mol
MDL Number: MFCD00150789
CAS Number: 18662-53-8
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty