Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chrome pure blue BX ~60% (dye content)

Supplier: Ambeed
Warning
Synonyms: Sunchromine Pure Blue BXMordant Blue 1Eriochrome Azurol B

A883274-5G
76729-024EA 20.6 USD InStock
76729-024
Chrome pure blue BX ~60% (dye content)
Chrome pure blue BX
Caution: for Research Use Only


Formula: C₂₃H₁₆Cl₂O₆
MW: 503.24 g/mol
MDL Number: MFCD00036446
CAS Number: 1796-92-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty