Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Mercaptomenthone 98%

Supplier: Ambeed
Danger
Synonyms: 8-Mercaptomenthonep-Mentha-8-thiol-3-one2-(1-Mercapto-1-methylethyl)-5-methylcyclohexanone

A891566-1G A891566-25G
76729-060EA 9.27 USD InStock
76729-060 76729-062
8-Mercaptomenthone 98%
8-Mercaptomenthone
Caution: for Research Use Only


Formula: C₁₀H₁₈OS
MW: 186.31 g/mol
MDL Number: MFCD00012393
CAS Number: 38462-22-5
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty