Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dipotassium hexachloropalladate 98%

Supplier: Ambeed
Warning

A927526-1G A927526-5G A927526-250MG A927526-100MG
76729-214EA 21.63 USD InStock
76729-214 76729-220 76729-216 76729-218
Dipotassium hexachloropalladate 98%
Dipotassium hexachloropalladate
Caution: for Research Use Only


Formula: Cl₆K₂Pd
MW: 397.33 g/mol
MDL Number: MFCD00043081
CAS Number: 16919-73-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty