Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-3-nitrobenzophenone 97%

Supplier: Ambeed
Warning

A955499-100G A955499-500G A955499-5G A955499-25G
76729-332EA 70.04 USD InStock
76729-332 76729-334 76729-336 76729-338
4-Chloro-3-nitrobenzophenone 97%
4-Chloro-3-nitrobenzophenone
Caution: for Research Use Only


Formula: C₁₃H₈ClNO₃
MW: 261.66 g/mol
MDL Number: MFCD00007081
CAS Number: 56107-02-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty