Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyacrylonitrile, M.W. ∼150,000

Supplier: Ambeed
Warning

A971305-25G A971305-100G
76729-362EA 19.57 USD InStock
76729-362 76729-364
Polyacrylonitrile, M.W. ∼150,000
Polyacrylonitrile
Caution: for Research Use Only


Formula: (C₃H₃N)n
MDL Number: MFCD00084395
CAS Number: 25014-41-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty