Would you like to log in again?
Eisco Premium Qualitative Filter Papers
Supplier: EISCO SCIENTIFIC LLC
Eisco
CH0390F CH0390C CH0390A CH0390B CH0390E CH0390D
470344-030EA 4.9 USD InStock
470344-030 470344-892 470345-232 470345-456 470344-982 470343-586
Eisco Premium Qualitative Filter Papers
Filters Filter Paper Qualitative Filter Paper
Pack of 100 premium Qualitative Filter Papers with medium speed (85 gsm) and 10 µm pore size.
  • High retentivity, perfect for general laboratory filtering and qualitative analysis
  • 10 µm (micron) pore size
  • Medium speed (85 gsm)
Premium quality filter papers for use in labs. Pack of 100 circles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty