Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Enhanced Clinical Centrifuge

Supplier: Globe Scientific
GCC-E50
76638-246EA 801.14 USD InStock
76638-246
Enhanced Clinical Centrifuge
Centrifuges Benchtop Centrifuges
For simple, reliable operation, compatible with a variety of tubes and with all the features needed without the cost of the ones not required, the GCC range of clinical centrifuges simply can’t be beaten.

  • Fast acceleration and deceleration
  • Speed range of 300 to 4000 rpm
  • Independent control of speed and time
  • Control by RCF
  • Suitable for numerous clinical and research applications, including separations
  • Compatible with 50 ml centrifuge tubes
  • GCC-E50 includes fixed angle rotor that holds up to six (6) 50 ml tubes or six (6) 15 ml tubes

GCC-E50 Enhanced Clinical Centrifuge is an economical solution for 15 and 50 ml separations with an enhanced user interface, easy to read backlit LCD display, tight accuracy, and automated self-diagnostics on startup. An extension based on the Globe Scientific GCC-E enhanced clinical centrifuge, the GCC-E50 employs a new rotor for spinning up to 6×50 ml centrifuge tubes or 6×15 ml centrifuge tubes. Programming allows for setting either by speed or G-force. A quick spin function also enables easy spin downs when a timed program isn’t necessary. The added safety of an electric locked lid ensures users don’t open the lid too soon after a protocol is completed.

Speed accuracy: ±20 rpm.

Certifications: CE, IVD.

Delivery information: Supplied with fixed angle rotor that holds up to 6×50 ml tubes or 6×15 ml tubes.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty