Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Portable Clinical Centrifuge

Supplier: Globe Scientific
GCC-P
76645-730EA 485.86 USD InStock
76645-730
Portable Clinical Centrifuge
Centrifuges Benchtop Centrifuges
The globe scientific GCC-P portable clinical centrifuge is designed for easy transportation to facilitate convenient field separations using the included 12 VDC car power adapter.

  • Operates using an included 12 VDC car adaptor (wall adapters sold separately)
  • Fast acceleration and two deceleration speeds
  • Speed range of 300 to 5000 rpm
  • Independent control of speed or RCF and time
  • Audible alert function
  • Ideal for use with numerous tube types
  • Includes fixed angle rotor that holds up to 6×15 ml centrifuge tubes or 6×1.5/2.0, 5, 7 or 10 ml tubes when adapters are used

The GCC-P Portable Centrifuge is compact in size and packed with the features required for routine clinical applications. The included six-position rotor accommodates 1.5/2.0 to 5, 7, 10 and 15 ml tubes (all adapters are included). A weighted base ensures the unit remains stable in field environments. An easy-to-read LCD display shows both time and speed or RCF. Two program buttons enable simple programing and recall of regularly used methods. The timer will not count down until the set speed is reached to ensure your separations and experiments work as intended. Safety is maintained by an automatic lid lock that only disengages when the program is completed or manually once the unit has completely stopped.

Speed accuracy: ±20 rpm.

Delivery information: Supplied with fixed angle rotor that holds 6×1.5/2.0, 5, 7, 10 and 15 ml tubes and a 12 VDC car adaptor.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty