Would you like to log in again?
FLIPR® Calcium 5 Fluorescent Assay Kit
Supplier: Molecular Devices
FLIPR®
R8185 R8186
MSPP-R8186EA 5531.63 USD InStock
MSPP-R8186 MSPP-R8185
FLIPR® Calcium 5 Fluorescent Assay Kit
Assays Cellular Assays
The FLIPR® Calcium 5 Assay Kit provides a fast, simple, and reliable fluorescence-based assay for detecting changes in intracellular calcium.
  • Maximises fluorescent signal by reducing cell wash artifacts
  • Reduced well-to-well variability, improving assay quality (Z' factor) and reliability (CV%) of high-throughput screens
  • Mix-and-read protocol accelerates assay workflow and increases throughput
  • Superior signal-to-noise ratio facilitates confirmation of endogenous or transiently transfected receptor activity during assay development
  • Pre-optimised and validated protocols for the FLIPR® Tetra and FlexStation® Systems ensure navigation to both routine and unconventional cell lines and targets
FLIPR® Calcium 5 Assay Kit contains a novel fluorophore that amplifies the assay signal window while leveraging the superior masking technology used in the FLIPR® Calcium 4 Assay Kit to reduce background fluorescence. The kit is designed to work for the majority of GPCRs, including Chemokine and other difficult receptors, sticky compounds, allosteric modulators, as well as with calcium channels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty