Would you like to log in again?
Solar Panel
Supplier: NEWARK ELECTRONICS
31AC4412
470350-854EA 51.4 USD InStock
470350-854
Solar Panel
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Solar panels feature a 2.5 W power rating and 12 V open circuit voltage.
  • 2.5 W solar battery charger panel
  • 12 V DC plug and alligator battery clamps included
  • Durable ABS plastic housing with mounting holes
  • Includes 4 suction cups for easy mounting to the windshield
  • 9' total cord length (1' from connector to adapter plug and 8' from adapter plug to solar panel)
Solar panel for all your off-the-grid 12 V battery charging and maintaining needs. Amorphous solar panels operate in all daylight conditions, even cloudy days.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty