Would you like to log in again?
Solar Cells, Monocrystalline
Supplier: NEWARK ELECTRONICS
94W9305 94W9302
470350-856EA 33.35 USD InStock
470350-856 470350-852
Solar Cells, Monocrystalline
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Maximum efficiency monocrystalline solar cells.
  • Encapsulated with low reflecting water clear epoxy to protect against physical impact and harsh outdoor environments
  • Terminated with a DC barrel plug Calculating Watts Power [Watt] = Voltage [Volt] Current [Ampere]
For applications requiring a higher voltage supply source, you can wire multiple solar panels in a series. Wire the positive terminal of solar panel #1 to the load, and connecting the negative terminal of solar panel #1 to the positive terminal of solar panel #2. In this wiring, the total voltage delivered to the load will be doubled and the rated current supply will remain the same.
If an application needs more power, wire multiple solar panels in parallel. By connecting the positive terminal of solar panel #1 to the postive terminal of solar panel #2, and connecting the negative terminal of solar panel #1 to the negative terminal of solar panel #2, the maximum current that can be delivered to the load will be doubled. The rated voltage will remain the same.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty