Would you like to log in again?
Magnetic Stand
Supplier: G-Biosciences
786-888
76285-444EA 346.86 USD InStock
76285-444
Magnetic Stand
Support Stands
Magnetic stand is designed for the efficent pull down of magnetic beads in 1.5 and 2 ml micro centrifgue tubes.
  • Suitable for 8×1.5 to 2 ml tubes
  • Pulls beads to tube wall to allow removal of all supernatant
The powerful magnet pulls beads to the side of the tube, allowing for the removal of all the liquid supernatant with no contamination with beads. It can also be used for immunoprecipitations and protein purification analysis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty