Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-D-phenylalanine 95%

Supplier: Ambeed
Warning
Synonyms: N-Ac-D-Phe-OHAc-D-Phe-OH

A104280-5G A104280-10G A104280-25G
77570-256EA 6 USD InStock
77570-256 76765-362 76765-360
N-Acetyl-D-phenylalanine 95%
N-Acetyl-D-phenylalanine
Caution: for Research Use Only


Formula: C₁₁H₁₃NO₃
MW: 207.23 g/mol
MDL Number: MFCD00002664
CAS Number: 10172-89-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty