Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-2-chlorobenzoic acid 98%

Supplier: Ambeed
Warning

A115385-1G A115385-10G A115385-100G A115385-5G
76766-134EA 5.15 USD InStock
76766-134 76766-136 76766-130 76766-132
4-Amino-2-chlorobenzoic acid 98%
4-Amino-2-chlorobenzoic acid
Caution: for Research Use Only


Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD00007772
CAS Number: 2457-76-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty