Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methyl-1,3-benzothiazol-6-amine 98%

Supplier: Ambeed
Warning
Synonyms: 6-Amino-2-methyl-1,3-benzothiazole

A132595-250MG A132595-1G A132595-5G
76767-298EA 53.56 USD InStock
76767-298 76767-300 76767-302
2-Methyl-1,3-benzothiazol-6-amine 98%
2-Methyl-1,3-benzothiazol-6-amine
Caution: for Research Use Only


Formula: C₈H₈N₂S
MW: 164.23 g/mol
MDL Number: MFCD00160076
CAS Number: 2941-62-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty